missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)amine, 99%

Product Code. 10518283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10518283 25 g Glass bottle 25g
10311903 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10518283 Supplier Thermo Scientific Acros Supplier No. 378770250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1586-73-8 | C9H27NSi3 | 233.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1586-73-8
Molecular Formula C9H27NSi3
Molecular Weight (g/mol) 233.58
MDL Number MFCD00047990
InChI Key PEGHITPVRNZWSI-UHFFFAOYSA-N
PubChem CID 74110
IUPAC Name [[bis(trimethylsilyl)amino]-dimethylsilyl]methane
SMILES C[Si](C)(C)N([Si](C)(C)C)[Si](C)(C)C

Specifications

Melting Point 67°C to 69°C
Color Colorless
Boiling Point 72°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula N(Si(CH3)3)3
Quantity 25 g
Synonym tris trimethylsilyl amine, amine, tris trimethylsilyl, silanamine, 1,1,1-trimethyl-n,n-bis trimethylsilyl, silanamine,1,1,1-trimethyl-n,n-bis trimethylsilyl, nitrilotris trimethylsilane, ch3 3si 3n, n,n,n-tris trimethylsilyl amine #, bis trimethylsilyl amino-dimethylsilyl methane
Solubility Information Solubility in water: insoluble
Formula Weight 233.58
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Tris(trimethylsilyl)amine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.