missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)amine, 99%, Thermo Scientific Chemicals

Product Code. 10311903
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1586-73-8
Molecular Formula C9H27NSi3
Molecular Weight (g/mol) 233.58
MDL Number MFCD00047990
InChI Key PEGHITPVRNZWSI-UHFFFAOYSA-N
PubChem CID 74110
IUPAC Name [[bis(trimethylsilyl)amino]-dimethylsilyl]methane
SMILES C[Si](C)(C)N([Si](C)(C)C)[Si](C)(C)C

Specifications

Melting Point 67°C to 69°C
Color Colorless
Boiling Point 72°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula N(Si(CH3)3)3
Quantity 5 g
Synonym tris trimethylsilyl amine, amine, tris trimethylsilyl, silanamine, 1,1,1-trimethyl-n,n-bis trimethylsilyl, silanamine,1,1,1-trimethyl-n,n-bis trimethylsilyl, nitrilotris trimethylsilane, ch3 3si 3n, n,n,n-tris trimethylsilyl amine #, bis trimethylsilyl amino-dimethylsilyl methane
Solubility Information Solubility in water: insoluble
Formula Weight 233.58
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Tris(trimethylsilyl)amine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'