missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsiloxy)silane, 97%

Product Code. 11399325
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399325 5 g 5g
11309335 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399325 Supplier Thermo Scientific Alfa Aesar Supplier No. L16686.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1873-89-8 | C9H27O3Si4 | 295.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1873-89-8
Molecular Formula C9H27O3Si4
Molecular Weight (g/mol) 295.65
MDL Number MFCD00054865
InChI Key XAASNKQYFKTYTR-UHFFFAOYSA-N
Synonym tris trimethylsiloxy silane, tris trimethylsilyloxy silane, tris trimethylsilyloxy silicon, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, 1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, tris trimethylsilyloxy silyl radical, trisiloxane, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy
PubChem CID 6365044
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 64°C (10mmHg)
Quantity 5 g
UN Number UN1993
Flash Point 50°C (122°F)
IUPAC Name 2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Formula Weight 296.66
Percent Purity 97%
Refractive Index 1.3865
Density 0.852
Chemical Name or Material Tris(trimethylsiloxy)silane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.