Learn More
Thermo Scientific Chemicals TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8.3
Description
Tris Tricine Gel Electrophoresis is routinely used for the separation of small proteins and peptides with a molecular weight of I to 100 kDa. It is used for protein analysis. This type of gel is perfectly suited for the separation and identification of proteins in samples containing only a few different proteins to samples containing complex mixtures (e.g. from cell extracts).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | TRIS-Tricine-SDS running buffer (10X) |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Plastic bottle |
| Solubility Information | It is soluble in water. |
Safety and Handling
- TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8.3
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]glycine, 2-amino-2-(hydroxymethyl)-1,3-propanediol, Sodium
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.