missing translation for 'onlineSavingsMsg'
Learn More

Tris, PlusOne™, Cytiva

Product Code. 11535005 Shop All Cytiva Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11535005

Brand: Life Science products Cytiva 17132101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for 2-D electrophoresis. Tris, PlusOne™, Cytiva is suitable for preparing electrophoresis buffer.

  • Qualified for 2-D electrophoresis
  • Tris (tris[hydroxymethyl] aminoethane)
  • Prepares buffer with pH range of 7.2 to 9.0.
  • Used in PAGE and Sequencing
  • Soluble in cold water and hot water, methanol
  • Solid physical form
TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1
Quantity 500 g
Assay Percent Range 99.8 to 100.1%
Molecular Formula C4H11NO3
Synonym Tris(hydroxymethyl)aminomethane
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.136
PubChem CID 6503
ChEBI CHEBI:9754
Formula Weight 121.14
Chemical Name or Material Tris, Suitable for preparing electrophoresis buffer in the range 7.2 to 9.0
Show More Show Less
compliance-icons
Signal Word
  • ||Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.