missing translation for 'onlineSavingsMsg'
Learn More

Tris(pentafluorophenyl)phosphine, 95%

Product Code. 11347825
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347825 5 g 5g
11337825 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347825 Supplier Thermo Scientific Alfa Aesar Supplier No. L02748.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1259-35-4 | C18F15P | 532.15 g/mol

Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Handle under inert gas. Incompatible with Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1259-35-4
Molecular Formula C18F15P
Molecular Weight (g/mol) 532.15
MDL Number MFCD00079654
InChI Key FQLSDFNKTNBQLC-UHFFFAOYSA-N
Synonym tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane
PubChem CID 102076
IUPAC Name tris(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 115°C to 119°C
Odor Odorless
Quantity 5 g
Beilstein 773243
Solubility Information Insoluble in water.
Formula Weight 532.15
Percent Purity 97%
Chemical Name or Material Tris(pentafluorophenyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.