missing translation for 'onlineSavingsMsg'
Learn More

Tris(pentafluorophenyl)borane, 97%

Product Code. p-7047215
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394885 1 g 1g
11304895 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394885 Supplier Thermo Scientific Alfa Aesar Supplier No. L18054.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1109-15-5 | C18BF15 | 511.98 g/mol

Tris(pentafluorophenyl)borane is used as an excellent activator component in homogeneous Ziegler-Natta chemistry. It is also used as a catalyst for reductions, alkylations, hydrometallation reactions and catalyzed aldol-type reactions. It is a useful Lewis acid and catalyzes hydrosilylation of aldehydes. Further, it is used in olefin polymerization catalysis. It serves as a reagent in the preparation of organometallic complexes. In addition to this, it is useful as polymerization catalysts and used to study the heterolytic cleavage of dihydrogen in associated with tri-tert-butylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(pentafluorophenyl)borane is used as an excellent activator component in homogeneous Ziegler-Natta chemistry. It is also used as a catalyst for reductions, alkylations, hydrometallation reactions and catalyzed aldol-type reactions. It is a useful Lewis acid and catalyzes hydrosilylation of aldehydes. Further, it is used in olefin polymerization catalysis. It serves as a reagent in the preparation of organometallic complexes. In addition to this, it is useful as polymerization catalysts and used to study the heterolytic cleavage of dihydrogen in associated with tri-tert-butylphosphine.

Solubility
Soluble in hexane, chloroform, dichloromethane, toluene, and polar solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and trimethylaluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1109-15-5
Molecular Formula C18BF15
Molecular Weight (g/mol) 511.98
MDL Number MFCD00269813
InChI Key OBAJXDYVZBHCGT-UHFFFAOYSA-N
Synonym tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
PubChem CID 582056
IUPAC Name tris(2,3,4,5,6-pentafluorophenyl)borane
SMILES FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 126°C to 130°C
Quantity 1 g
UN Number UN2811
Beilstein 2931347
Sensitivity Moisture sensitive
Merck Index 14,9755
Solubility Information Soluble in hexane,chloroform,dichloromethane,toluene,and polar solvents.
Formula Weight 511.99
Percent Purity 97%
Chemical Name or Material Tris(pentafluorophenyl)borane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.