missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]yttrium(III), 98+%, Thermo Scientific Chemicals

Product Code. 11344487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344487 5 g 5g
11334487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344487 Supplier Thermo Scientific Alfa Aesar Supplier No. 039578.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41836-28-6
Molecular Formula C18H57N3Si6Y
Molecular Weight (g/mol) 573.091
MDL Number MFCD00210649
InChI Key WJMJCIJUJNENIA-UHFFFAOYSA-N
Synonym tris bis trimethylsilyl amido yttrium
PubChem CID 131853833
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane;yttrium
SMILES C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C.[Y]

Specifications

Melting Point 180°C to 184°C
Color White
Boiling Point 105°C (10-4mmHg)(sublimation)
Packaging Ampule
Linear Formula [[(CH3)3Si]2N]3Y
Quantity 5 g
UN Number UN3396
Sensitivity Moisture Sensitive
Formula Weight 570.06
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis(trimethylsilyl)amide]yttrium(III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.