missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III), 98%

Product Code. 11408520
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408520 5 g 5g
11418520 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408520 Supplier Thermo Scientific Alfa Aesar Supplier No. 044359.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35789-03-8 | C18H57GdN3Si6 | 641.435 g/mol

Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III) is used in many asymmetric catalysis applications, glove box and Schlenk techniques should be employed to prevent exposure of the rare earth catalyst to air and moisture, which can be detrimental to the reaction outcome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35789-03-8
Molecular Formula C18H57GdN3Si6
Molecular Weight (g/mol) 641.435
MDL Number MFCD03427091
InChI Key SDYADEGNUOTJQY-UHFFFAOYSA-N
Synonym tris n n-bis trimethylsilyl amide gado
PubChem CID 131878399
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane;gadolinium
SMILES C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C.C[Si](C)(C)N[Si](C)(C)C.[Gd]

Specifications

Melting Point 161°C to 167°C
Linear Formula [[(CH3)3Si]2N]3Gd
Quantity 5 g
UN Number UN3396
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 638.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]gadolinium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.