Learn More
Tris[N,N-bis(trimethylsilyl)amide]cerium(III), 98%, Thermo Scientific Chemicals
Description
Tris[N,N-bis(trimethylsilyl)amide]cerium(III) is used as a reactant for synthesis of amidocerium(IV) oxides, Sterically encumbered N-heterocyclic carbene ligands, Lanthanide imidodiphosphinate complexes with electroluminescent properties, Lanthanide complexes of an oxazoline-phenoxide hybrid chelate. Some of the other applications include it is used as a reactant for One-electron oxidation to produce tetravalent cerium compounds, Addition-elimination reactions across the M-C bond of metal N-heterocyclic carbenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Linear Formula | [[(CH3)3Si]2N]3Ce |
| Quantity | 1 g |
| UN Number | UN3396 |
| Sensitivity | Moisture Sensitive |
| Formula Weight | 621.27 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Tris[N,N-bis (trimethylsilyl)amide]cerium (III) |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.