missing translation for 'onlineSavingsMsg'
Learn More

Tris, MP Biomedicals™

Product Code. 11483541 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
This item is not returnable. View return policy

Product Code. 11483541

Brand: MP Biomedicals 02103133.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful as a buffer in a variety of biological systems. Tris, MP Biomedicals is used for pH control of body fluids and in buffering systems for electrophoresis. It is ultra-pure, RNAse and DNAse-free, and relatively non-hygroscopic. It can be dried at 100°

Tris have been useful as buffers in a wide variety of biological systems. It is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.

  • Used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions
  • Used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes)
  • Does not precipitate calcium salts and is of value in maintaining solubility of manganese salts
  • Can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol, Tromethamine, Tris-[hydroxymethyl]amino-methane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL).
Formula Weight 121.14
Percent Purity >99.9%
Grade Ultrapure
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.