missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Tris may be used by itself as a buffer or as a component of mixed buffer formulations e.g. reaction buffers for restriction enzymes or nucleic acid modifying enzymes.
- Tris salts are used in protein crystallization at various pH values.
- The use of low-ionic strength Tris buffers in the formation of intermediate filaments of lamin from Caenorhabditis elegans has been described.
- Tris has been utilized in studies of double stranded complexes of peptide nucleic acids (PNA) and their complementary DNA sequences.
- The use of Tris in capillary electrochromatography and UV analysis of tocopherols and tocotrienols has been reported.
- Tris is an established basimetric standard and buffer used in biochemistry and molecular biology
Specifications
Specifications
| Melting Point | 167°C to 172°C (literature) |
| Boiling Point | 219°C to 220°C/10 mmHg(literature) |
| Color | White |
| pH | 10.5 to 11.5 (0.5 M aq. soln., 20°C) |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Beilstein | 741883 |
| Solubility Information | Soluble in water (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), chloroform (0.05mg/mL), and alkaline solution. |
| Grade | >99.8% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?