missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, Ultra Pure, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Description
The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per°C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Solubility Information | Solubility (5% aq soln) Clear, Colorless Solution. |
| Formula Weight | 157.6g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction