missing translation for 'onlineSavingsMsg'
Learn More

Tris Hydrochloride, MP Biomedicals™

Product Code. 11463531 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463531 500 g 500g
11443531 100 g 100g
11453531 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463531 Supplier MP Biomedicals Supplier No. 02103130.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful as buffers in a wide variety of biological systems. Tris Hydrochloride, MP Biomedicals provides pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications. It has been used as a starting material for p

Uses include:

  • pH control in vitro and in vivo for body fluids
  • In buffering systems for electrophoresis applications
  • As a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes)
  • Direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Trometamol, Tromethamine, Tris-[hydroxymethyl]aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL)
Formula Weight 157.6
Percent Purity >99%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.