missing translation for 'onlineSavingsMsg'
Learn More

TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™

Product Code. 11494922 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494922 1 kg 1kg
11474922 100 g 100g
11484922 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494922 Supplier MP Biomedicals Supplier No. 0219455891

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Excellent biochemical and biological buffer for all applications where ultra high purity is required. The pH values of all buffers are temperature and concentration dependent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range > 99%
Formula Weight 157.6
Grade Cell Culture
Chemical Name or Material TRIS hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.