missing translation for 'onlineSavingsMsg'
Learn More

Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™

Product Code. 10336763
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336763 100 mL Poly Bottle 100mL
10795104 500 mL Poly Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336763 Supplier Fisher BioReagents Supplier No. BP1758100
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.)

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 8.0 provide convenient and highly purified solutions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1 , 1185-53-1 , 7732-18-5
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris-HCl
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Color Undesignated
Concentration 0.96 to 1.04 M
pH 10
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Identification Pass Test
Packaging Poly Bottle
Protease Protease free
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Hydrochloride, 1M Solution, pH 8.0
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.