Learn More
Tris-Glycine-SDS, 10X Powder (Electrophoresis), Fisher BioReagents
Brand: Fisher Bioreagents BP1342-1
Code : 32
Additional Details : Weight : 1.00000kg
Description
Tris-Glycine-SDS is commonly used as a buffer in SDS-PAGE.Specifications
Tris-Glycine-SDS | |
77-86-1,56-40-6,151-21-3 | |
Solid | |
Foil Pack | |
Electrophoresis |
10X Powder | |
White | |
1 Pk. | |
Protease free |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tris Glycine SDS 10X Powder
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.