missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)cobalt chloride trihydrate, 97%, Thermo Scientific™

Product Code. 15520604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15520604 25 g 25g
15510604 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15520604 Supplier Thermo Scientific Acros Supplier No. 458960250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14883-80-8
Molecular Formula C6H24Cl3CoN6·3H2O
Molecular Weight (g/mol) 399.64
InChI Key WBMVTRUQSMZIKW-UHFFFAOYSA-K
Synonym tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
PubChem CID 73995044
IUPAC Name cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
SMILES C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]

Specifications

Melting Point >300.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 399.64
Percent Purity 97%
Chemical Name or Material Tris(ethylenediamine)cobalt chloride trihydrate
compliance-icons
Product Identifier
  • Tris(ethylenediamine)cobalt chloride trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.