missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 1X Solution, pH 7.6, Molecular Biology, Fisher BioReagents

Product Code. 10204973
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204973 100 mL Poly Bottle 100mL
10183293 500 mL Poly Bottle 500mL
10173293 1 L Nalgene Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10204973 Supplier Fisher BioReagents Supplier No. BP2474100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10mM tris and 1mM EDTA
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-EDTA
Name Note 1X Solution, pH 7.6
CAS 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Filtered Through Filtered through a 5-micron filter.
Packaging Poly Bottle
Protease Protease free
Synonym TE
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.