missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TE Buffer, Tris-EDTA, 1X Solution, pH 7.6, Molecular Biology, Fisher BioReagents
Description
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
- Eliminates the hassle of dilution or waiting for powder to dissolve
- 1X solution contains 10mM tris and 1mM EDTA
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- TE buffer is routinely used for resuspending nucleic acid samples
- Intended for laboratory research use only
Specifications
Specifications
| Chemical Name or Material | Tris-EDTA |
| Name Note | 1X Solution, pH 7.6 |
| CAS | 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5 |
| Color | Colorless |
| pH | 7.4 to 8.1 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| DNase | DNase free |
| Filtered Through | Filtered through a 5-micron filter. |
| Packaging | Poly Bottle |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?