missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 100X Powder, pH 8.0, Molecular Biology, Fisher BioReagents

Product Code. 10511255
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10511255

Brand: Fisher BioReagents BP13391

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Save time and simplify your preparation of buffer concentrate by using Fisher BioReagents 100X TE powder - simply add pure water
  • Each pack contains the correct amount of pre-weighed powder to make 1L of 100X TE buffer containing 1.0 M tris and 0.1M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-EDTA
Name Note 100X Powder
CAS 77-86-1, 60-00-4
Color White
Physical Form Solid
Quantity 1 Pk.
DNase DNase free
Packaging Foil Pack
Protease Protease free
Synonym TE
Solubility Information Soluble in water
Grade Molecular Biology
Show More Show Less
compliance-icons
Product Identifier
  • Tris EDTA 100X Powder
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.