missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)methane, 85%, ACROS Organics™

Brand:  Acros Organics 205700050

Code : 29

Additional Details : CAS Number : 5762-56-1 Weight : 0.05500kg Transport : UN number : 2733 Chem class : 1b Pack group : II


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10331803

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Tris(dimethylamino)methane
85%
CH[N(CH3)2]3
MFCD00008322
≥80%
C7H19N3
145.25

Specifications

Tris(dimethylamino)methane
Authentic
≥80%
C7H19N3
Glass bottle
MFCD00008322
Clear colorless to slightly yellow
5762-56-1
Liquid
85%
CH[N(CH3)2]3
145.25
0.8380g/mL
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tris(dimethylamino)methane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • H224-Extremely flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.

SDS