missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals

Product Code. 11329328
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Preparation of aluminum nitrides

Preparation of aluminum nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32093-39-3
Molecular Formula C12H36Al2N6
Molecular Weight (g/mol) 318.425
MDL Number MFCD00269811
InChI Key JGZUJELGSMSOID-UHFFFAOYSA-N
Synonym tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum
PubChem CID 16688802
IUPAC Name N-[bis(dimethylamino)alumanyl]-N-methylmethanamine
SMILES CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C

Specifications

Melting Point 87°C to 89°C
Color Brown to White
Density 0.865 g/mL
Boiling Point 90°C (0.05 mmHg)(sublimation)
Flash Point 21°C (70°F)
Linear Formula Al2[N(CH3)2]6
Quantity 1 g
UN Number UN3131
Sensitivity Moisture sensitive
Formula Weight 318.42
Physical Form Powder
Chemical Name or Material Tris (dimethylamino)aluminum dimer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(dimethylamino)aluminum dimer
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H280-Contains gas under pressure; may explode if heated.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'