missing translation for 'onlineSavingsMsg'
Learn More

Tris(diethylamino)phosphine, TRC

Product Code. 30450521
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450521 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450521 Supplier Toronto Research Chemicals Supplier No. TRCT8845305G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(diethylamino)phosphine
CAS 2283-11-6
Molecular Formula C12H30N3P
Quantity 5 g
Synonym N,N,N',N',N'',N''-Hexaethylphosphorous Triamide, Hexaethylphosphorous Triamide, Hexaethyltriamidophosphite, Hexaethyltriaminophosphine, Phosphorous Acid Tris(diethylamide), Tris(N,N-diethylamino)phosphine, Tris(diethylamido)phosphine
InChI Formula InChI=1S/C12H30N3P/c1-7-13(8-2)16(14(9-3)10-4)15(11-5)12-6/h7-12H2,1-6H3
SMILES CCN(CC)P(N(CC)CC)N(CC)CC
IUPAC Name N-[bis(diethylamino)phosphanyl]-N-ethylethanamine
Molecular Weight (g/mol) 247.36
Formula Weight 247.2177
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.