missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%

Product Code. 10621663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10621663 1 g Glass bottle 1g
11964361 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10621663 Supplier Thermo Scientific Acros Supplier No. 369340010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52522-40-4 | C52H43Cl3O3Pd2 | 1035.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52522-40-4
Molecular Formula C52H43Cl3O3Pd2
Molecular Weight (g/mol) 1035.10
MDL Number MFCD00075479
InChI Key LNAMMBFJMYMQTO-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
PubChem CID 11029508
IUPAC Name chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 131.0°C to 135.0°C
Color Black to Purple
Assay Percent Range 96% min. (ex Pd)
Packaging Glass bottle
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2·CHCl3
Quantity 1 g
Fieser 14,339; 15,353; 16,372; 17,394
Formula Weight 1035.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tris(dibenzylideneacetone)dipalladium-chloroform

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.