missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform, Pd 20.6%, Thermo Scientific Chemicals

£84.80 - £347.00

Chemical Identifiers

CAS 52522-40-4
Molecular Formula C52H43Cl3O3Pd2
Molecular Weight (g/mol) 1035.10
MDL Number MFCD00075479
InChI Key LNAMMBFJMYMQTO-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
PubChem CID 11029508
IUPAC Name chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15416655
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044969.03
1 g
£84.80
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15426655
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044969.06
5 g
£347.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tris(dibenzylideneacetone)dipalladium(0) is used in Synthesis of nanosized palladium phosphides upon interaction with white phosphorous, Preparation of palladium triphenylphosphine carbonyl cluster complexes, Precursor for synthesis of functionalized multiwalled carbon nanotube-palladium complexes used as catalysts for Heck coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52522-40-4
1035.10
LNAMMBFJMYMQTO-UHFFFAOYSA-N
11029508
[Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
C52H43Cl3O3Pd2
MFCD00075479
tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium

Specifications

52522-40-4
C52H43Cl3O3Pd2
MFCD00075479
tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
LNAMMBFJMYMQTO-UHFFFAOYSA-N
chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
11029508
Pd 20.6%
Tris (dibenzylideneacetone)dipalladium (0), Complex with chloroform
131°C to 135°C
Pd2(C17H14O)3·CHCl3
Air and moisture sensitive
Soluble in organic solvents. Insoluble in water.
[Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
1035.10
1035.11
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H373-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.

P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H320-H351

missing translation for 'tsca' : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only