missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(cyclopentadienyl)yttrium(III), 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
Tris(cyclopentadienyl)yttrium(III) is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Melting Point | 295°C |
Boiling Point | 200°C (sublimation)(vacuum) |
Packaging | Ampule |
Quantity | 0.5 g |
UN Number | UN3181 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 284.2 |
Percent Purity | 99.9% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'