missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)lanthanum(III), 99.9% (REO), Thermo Scientific™

Product Code. 11353648
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353648 1 g 1g
11363648 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353648 Supplier Thermo Scientific Alfa Aesar Supplier No. 039251.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1272-23-7
Molecular Formula C15H15La
Molecular Weight (g/mol) 334.19
MDL Number MFCD00045467
InChI Key WMTALVRZNMWNTJ-UHFFFAOYSA-N
Synonym lanthanum 3+ ion tris cyclopenta-1,3-dien-1-ide
PubChem CID 22481131
IUPAC Name cyclopenta-1,3-diene;lanthanum(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[La+3]

Specifications

Melting Point 295°C (decomposition)
Packaging Ampule
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 334.19
Percent Purity 99.9%
Physical Form Crystalline
Assay (REO)
Chemical Name or Material Tris(cyclopentadienyl)lanthanum(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.