missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ TRIS-CAPS Buffer, 10X

For blot transfer

Brand:  Alfa Aesar™ J61339.K2

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15423949

  • £67.80 / 1 L


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Specifications

1L
Liquid
Buffer
For blot transfer
10X
Videos
SDS