missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X) pH 7.4
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
Description
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.4.
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.4.
Specifications
Specifications
| Concentration | 10X |
| pH | 7.4 |
| Physical Form | Liquid |
| Quantity | 2 L |
| Odor | Odorless |
| Solubility Information | Soluble in water and organic solvents. |
| Chemical Name or Material | TRIS-buffered saline |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction