Learn More
TRIS-buffered saline (TBS, 10X), with 1% Triton™ X-100, Thermo Scientific Chemicals
Description
It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.
Solubility
Fully miscible in water.
Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4
Specifications
Specifications
| Concentration | 10X |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solubility Information | Fully miscible in water. |
| Chemical Name or Material | TRIS-buffered saline, with 1% Triton™ X-100 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.