missing translation for 'onlineSavingsMsg'
Learn More

Tris Buffer 50 mM, Lonza™

Product Code. 11680441
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11680441 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11680441 Supplier Lonza Supplier No. S50642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used in place of water as the sample diluent for highly acidic or basic samples.

  • Can be used in place of water as the sample diluent for highly acidic or basic samples (for endotoxin testing, samples and sample dilutions should be between pH 6-8).
  • May be used to prepare samples for any of our endotoxin detection assays.
  • Contains <0.005 EU/mL endotoxin.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Buffer
Color Clear
Description Contains <0.005 EU/mL endotoxin
Concentration 50 mM
pH 8
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.