missing translation for 'onlineSavingsMsg'
Learn More

Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents

Product Code. 10705114
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10705114

Brand: Fisher BioReagents BP1759500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 2M, provides a convenient stock solution for preparing Tris buffers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Color Undesignated
Concentration 1.95 to 2.05 M
Physical Form Liquid
Quantity 500 mL
DNase DNase free
Identification Pass Test
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 9951
Grade Molecular Biology
Chemical Name or Material Tris Buffer, 2M Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.