missing translation for 'onlineSavingsMsg'
Learn More

Tris-Borate-Edta Buffer, for Electrophoresis, (10X Concentration), Fisher Chemical™

Product Code. 10204260 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 L
Packaging:
Bag in box
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204260 10 L Bag in box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204260 Supplier Fisher Chemical Supplier No. T/P050/24
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tris-Borate-Edta Buffer, for Electrophoresis, (10X Concentration)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610769-35-2
Molecular Formula C14H30BN3O14
Molecular Weight (g/mol) 475.211
MDL Number 4679
InChI Key OSBLTNPMIGYQGY-UHFFFAOYSA-N
PubChem CID 21257725
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;boric acid
SMILES B(O)(O)O.C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.C(C(CO)(CO)N)O

Specifications

Boiling Point 219°C
Color Colorless
pH 8.2 to 8.4
Physical Form Liquid
Quantity 10 L
Packaging Bag in box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.