missing translation for 'onlineSavingsMsg'
Learn More

Tris Borate EDTA Buffer

Product Code. 11538035
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11538035

Brand: Lonza 51234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 610769-35-2
Molecular Formula C14H30BN3O14
Quantity 1 L
InChI Key OSBLTNPMIGYQGY-UHFFFAOYSA-N
SMILES B(O)(O)O.C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;boric acid
Molecular Weight (g/mol) 475.211
PubChem CID 21257725
Chemical Name or Material Tris borate EDTA Buffer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.