missing translation for 'onlineSavingsMsg'
Learn More

Tris-(benzyltriazolylmethyl)amine, TRC

Product Code. 30451733
Change view
Click to view available options
Quantity:
25 mg
1 g
Unit Size:
1g
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451733 25 mg 25mg
30457304 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451733 Supplier Toronto Research Chemicals Supplier No. TRCT87629525MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 510758-28-8
Molecular Formula C30H30N10
Molecular Weight (g/mol) 530.63
Synonym 1-(1-Benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine, N,N,N-Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, TBTA, Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
IUPAC Name 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
SMILES C(N(Cc1cn(Cc2ccccc2)nn1)Cc3cn(Cc4ccccc4)nn3)c5cn(Cc6ccccc6)nn5

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C30H30N10/c1-4-10-25(11-5-1)16-38-22-28(31-34-38)19-37(20-29-23-39(35-32-29)17-26-12-6-2-7-13-26)21-30-24-40(36-33-30)18-27-14-8-3-9-15-27/h1-15,22-24H,16-21H2
Formula Weight 530.2655
Chemical Name or Material Tris-(benzyltriazolylmethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.