missing translation for 'onlineSavingsMsg'
Learn More

Melford Tris Base Ultra Pure

Product Code. 30018879
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
500g
5kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018879 100 g 100g
30018883 500 g 500g
30018880 1 kg 1kg
30018884 5 kg 5kg
30018882 25 kg 25kg
30018881 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30018879 Supplier Melford Supplier No. T60040100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tris Base Ultra Pure is a commonly used reagent and buffer component.

Tris Base Ultra Pure is a commonly used reagent and buffer component. Tris Base, Tris(hydroxymethyl)aminomethane, is a primary amine that acts as a buffering agent to maintain a stable pH in experimental procedures.

It is particularly effective in the pH range of 7.2 to 9.0, making it suitable for applications requiring slightly alkaline to mildly acidic pH conditions. Tris Base Ultra Pure is highly purified to remove impurities and contaminants, ensuring the highest quality for laboratory use.

The ultra-pure form is essential for applications where the presence of impurities can negatively impact experimental results. Tris Base Ultra Pure is commonly used in various molecular biology techniques such as DNA and RNA extraction, protein purification, and enzymatic assays.

It is also utilized in the preparation of electrophoresis buffers and PCR reactions.

Moisture: <0.2%

Color: <20 APHA (colorless)

Heavy Metals (as Pb): <0.0001%

Insoluble Matter: <0.005%

Heavy Metals: 5 ppm

pKa (20°C): 8.1 ± 0.1

Specifications

CAS 77-86-1
Quantity 100 g
Molecular Formula C4H11NO3
Grade Molecular Biology Ultrapure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.