missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TRIS base

Product Code. 18055442
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 18055442

Brand: Tocris Bioscience™ 3163/500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used laboratory reagent; used to make TBS, TAE and TBE buffers

Commonly used laboratory reagent. Highly water soluble. Working pH range 7 - 9. Used in buffer solutions such as Tris-buffered saline (TBS), Tris-acetate-ETDA (TAE) in combination with acetic acid and EDTA (Cat. No. 2811) and Tris-borate-EDTA in combination with boric acid and EDTA.
TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1
Quantity 500 g
Molecular Formula C4H11NO3
Formula Weight 121.14
Purity >99.9%
Molecular Weight (g/mol) 121.136
Chemical Name or Material TRIS base
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.