missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™

Product Code. 10326463
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
Unit Size:
1 L
20 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 10326463

Brand: Fisher BioReagents BP133220

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT

Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 50X TAE Solution - simply dilute as needed
  • 50X solution contains 2M tris-acetate and 0.050M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 50X TAE
  • Solution with 49 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 50X Solution
CAS 77-86-1, 60-00-4, 6850-28-8
Physical Form Liquid
Quantity 20 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly CUBE
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (24%), Acetic Acid (5.0%), and EDTA (<2%).
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.