missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents

Product Code. 10123293
Change view
Click to view available options
Quantity:
4 L
20 L
Unit Size:
20 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10123293 20 L 20 L
10542785 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123293 Supplier Fisher BioReagents Supplier No. BP243420
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TAE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 0.04M tris-acetate and 0.001M EDTA
  • Tested for the absence of DNase, RNase, and protease
  • Functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 1X Solution
CAS 77-86-1, 6850-28-8, 139-33-3, 10378-23-1, 7732-18-5
Color Undesignated
Physical Form Liquid
Quantity 20 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging Poly CUBE
Protease Protease free
Synonym TAE
Grade Electrophoresis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.