missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-acetate buffer solution, pH 8.5 EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
| Chemical Name or Material | Tris acetate buffer solution |
| pH | 8.5 |
| Quantity | 1000 mL |
| Packaging | Bottle |
| Grade | Eu. Ph. Reference |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction