missing translation for 'onlineSavingsMsg'
Learn More

Melford TRIS Acetate

Product Code. 30018904
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
30018904 500 g 500g
30018903 1 kg 1kg
30018902 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 30018904

Brand: Melford T60070500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Moisture: <0.3%

pH (0.1M, H2O) 25°C: 6.0-7.0%

Heavy Metals: 0.001%

Specifications

CAS 6850-28-8
Quantity 500 g
Molecular Formula C14H11NO3C2H4O2
Grade Ultra Pure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.