missing translation for 'onlineSavingsMsg'
Learn More

Tris, 99.0 to 101.0% (Dry Basis), Thermo Scientific Chemicals

Catalog Number p-7082837
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
Tris(hydroxymethyl)amino methane
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
Tris(hydroxymethyl)amino methane
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
Melting Point 171°C to 172°C
Boiling Point 219°C to 220°C (10 mmHg)
Physical Form Powder
Quantity 500 g
Linear Formula NH2C(CH2OH)3
Packaging Plastic bottle
Beilstein 741883
Formula Weight 121.14
Percent Purity 99.0 to 101.0%
Assay Dry basis
Chemical Name or Material Tris
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'