missing translation for 'onlineSavingsMsg'
Learn More

Tris, 99.0 to 101.0% (Dry Basis), Thermo Scientific™

Brand:  Thermo Scientific J22674.A7

Additional Details : CAS Number : 77-86-1 Weight : 5.00000kg

Packaging Plastic bottle
Quantity 5 kg

 View more versions of this product

Product Code. 15865348

  • £235.62 / 5kg
Estimated Shipment: 17-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
Tris(hydroxymethyl)amino methane
CHEBI:9754
C(C(CO)(CO)N)O

Specifications

Tris
77-86-1
NH2C(CH2OH)3
741883
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
219°C to 220°C (10 mmHg)
5 kg
Powder
C4H11NO3
MFCD00004679
Tris(hydroxymethyl)amino methane
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Plastic bottle
171°C to 172°C
Videos
SDS
Documents
Certificates
Special Offers

Special Offers

RUO – Research Use Only