missing translation for 'onlineSavingsMsg'
Learn More

Tris(8-hydroxyquinolinato)aluminum, 99%

Product Code. p-7052752
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15441968

Brand: Thermo Scientific Alfa Aesar H55656.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2085-33-8

Key green-light emitting and electron transport material for OLEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Key green-light emitting and electron transport material for OLEDs.

Solubility
Solubility in chloroform

Notes
Reacts with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2085-33-8
MDL Number MFCD00191693
Synonym tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane

Specifications

Melting Point >300°C
Color Yellow to Green
Quantity 1 g
Merck Index 14,370
Formula Weight 459.44
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tris(8-hydroxyquinolinato)aluminum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.