missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III), TRC

Product Code. 30451867
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451867 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451867 Supplier Toronto Research Chemicals Supplier No. TRCT794558500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
CAS 17631-68-4
Molecular Formula C30 H30 Eu F21 O6
Quantity 500 mg
Synonym Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')-Europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-κO,κO')-Europium, 3,5-Octanedione','6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-','europium complex Europium tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octadionate), Europium(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione), Tris(6,6,7,7,8,8,8-hetafluoro-2,2-dimethyl-3,5-octanedionato)europium','Tris(7,7-dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedionato)europium, NSC 379435, Sievers' reagent, Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionato)Eu(III), Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionato)europium, Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione)europium, Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione)europium(III), Tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyloctane-4,6-dionato)europium(III), Tris(2,2-dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedionato)europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III), Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')europium, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-κO,κO')europium, Tris(7,7-dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedionato)europium, Tris(perfluorobutanoylpivaloylmethano)europium
InChI Formula InChI=1S/3C10H10F7O2.Eu/c3*1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h3*4H,1-3H3;/q3*-1;+3
SMILES CC(C)(C)C1=[O][Eu+3]23([O]=C([CH-]C(=[O]2)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)([O]=C([CH-]C(=[O]3)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)[O]=C([CH-]1)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 1037.486
Formula Weight 1038.092
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.