missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)

Product Code. 11397748
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397748 5 g 5g
11307758 250 mg 250mg
11317758 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397748 Supplier Thermo Scientific Alfa Aesar Supplier No. 033541.06
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17631-68-4 | C30H33EuF21O6 | 1040.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17631-68-4
Molecular Formula C30H33EuF21O6
Molecular Weight (g/mol) 1040.52
MDL Number MFCD00064655,MFCD01074933
InChI Key UDXLMYFGTHAWDC-UHFFFAOYSA-N
Synonym eu fod 3, sievers' reagent, europium-fod, siever's reagent, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium iii, resolve-al tm eufod, chloride ionophore vii, tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato europium, europium iii-tris 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate
PubChem CID 6000041
IUPAC Name europium;(Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one
SMILES [Eu].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 203°C to 207°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 1037.49
Physical Form Powder
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.