missing translation for 'onlineSavingsMsg'
Learn More

Tris[4-(trifluoromethyl)phenyl]phosphine, 98%

Product Code. 11425333
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425333 5 g 5g
11415333 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425333 Supplier Thermo Scientific Alfa Aesar Supplier No. A12885.06
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13406-29-6 | C21H12F9P | 466.286 g/mol

Tris[4-(trifluoromethyl)phenyl]phosphine is a raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[4-(trifluoromethyl)phenyl]phosphine is a raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13406-29-6
Molecular Formula C21H12F9P
Molecular Weight (g/mol) 466.286
MDL Number MFCD00058883
InChI Key PXYCJKZSCDFXLR-UHFFFAOYSA-N
Synonym tris 4-trifluoromethylphenyl phosphine, tris 4-trifluoromethyl phenyl phosphine, tris 4-trifluoromethyl phenyl phosphane, phosphine, tris 4-trifluoromethyl phenyl, tris p-trifluoromethylphenyl phosphine, phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl, tris p-trifluoromethylphenyl phosphine,min, acmc-20ajbq
PubChem CID 139448
IUPAC Name tris[4-(trifluoromethyl)phenyl]phosphane
SMILES C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F

Specifications

Melting Point 72°C to 75°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 466.29
Percent Purity 98%
Chemical Name or Material Tris[4-(trifluoromethyl)phenyl]phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.