Learn More
Tris(4-fluorophenyl)phosphine, 98%
Description
Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 78°C to 82°C |
| Quantity | 25 g |
| Beilstein | 919838 |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 316.27 |
| Percent Purity | 98% |
| Chemical Name or Material | Tris(4-fluorophenyl)phosphine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.