missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-bromophenyl)amine, 98%, Thermo Scientific Chemicals

Product Code. 15407368
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407368 10 g 10g
15417368 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407368 Supplier Thermo Scientific Alfa Aesar Supplier No. H60860.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)

Solubility
Soluble in toluene.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4316-58-9
Molecular Formula C18H12Br3N
Molecular Weight (g/mol) 482.013
MDL Number MFCD00009665
InChI Key ZRXVCYGHAUGABY-UHFFFAOYSA-N
Synonym tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9
PubChem CID 258027
IUPAC Name 4-bromo-N,N-bis(4-bromophenyl)aniline
SMILES C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br

Specifications

Melting Point 141°C to 143°C
Quantity 10 g
Solubility Information Soluble in toluene.
Formula Weight 482.01
Percent Purity 98%
Chemical Name or Material Tris(4-bromophenyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.